Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN14
Entrez Gene ID
81619
Gene Name
tetraspanin 14
Gene Aliases
DC-TM4F2, TM4SF14
Chromosome Location
Chr.10: 80454271 - 80522635 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128309.1 NP_001121781.1 4 - 5 361 97
NM_030927.2 NP_112189.2 7 - 8 730 97
XM_006717992.3 XP_006718055.1 7 - 8 886 97
XM_011540219.2 XP_011538521.1 9 - 10 920 97
XM_011540220.2 XP_011538522.1 9 - 10 842 97
XM_011540221.2 XP_011538523.1 8 - 9 704 97
XM_011540222.2 XP_011538524.1 8 - 9 882 97
XM_017016707.1 XP_016872196.1 9 - 10 878 97
XM_017016708.1 XP_016872197.1 8 - 9 839 97
XM_017016709.1 XP_016872198.1 8 - 9 887 97
XM_017016710.1 XP_016872199.1 8 - 9 838 97
XM_017016711.1 XP_016872200.1 8 - 9 703 97
XM_017016712.1 XP_016872201.1 8 - 9 906 97
XM_017016713.1 XP_016872202.1 7 - 8 726 97
GenBank mRNA AF311903.1 - 7 - 8 695 97
AK295321.1 - 4 - 5 332 97
AL136638.1 - 7 - 8 701 97
BC002920.2 - 6 - 7 625 97
BC093016.1 - 7 - 8 730 97
BI918996.1 - 4 - 5 384 97
DC300676.1 - 4 - 5 333 97

View Full Product Details