Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRIP3
Entrez Gene ID
56675
Gene Name
nuclear receptor interacting protein 3
Gene Aliases
C11orf14, NY-SAR-105
Chromosome Location
Chr.11: 8980576 - 9004049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020645.2 NP_065696.1 4 - 5 672 103
GenBank mRNA AK024750.1 - 4 - 5 625 103
AK098694.1 - 4 - 5 625 103
AY211930.1 - 3 - 4 369 103
BC008835.2 - 4 - 5 608 103

View Full Product Details