The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164.4 | NP_001155.1 | 1 - 2 | 125 | 79 |
NR_047512.2 | 1 - 2 | 125 | 79 | ||
XM_006718211.1 | XP_006718274.1 | 1 - 2 | 366 | 79 | |
XM_006718213.1 | XP_006718276.1 | 1 - 2 | 366 | 79 | |
XM_017017639.1 | XP_016873128.1 | 1 - 2 | 366 | 79 | |
XM_017017640.1 | XP_016873129.1 | 1 - 2 | 366 | 79 | |
XM_017017641.1 | XP_016873130.1 | 1 - 2 | 366 | 79 | |
GenBank mRNA | AF394214.1 | - | 1 - 2 | 84 | 79 |
AK293452.1 | - | 1 - 2 | 82 | 79 | |
AK311725.1 | - | 1 - 2 | 85 | 79 | |
BG699532.1 | - | 1 - 2 | 125 | 79 | |
L77864.1 | - | 1 - 2 | 84 | 79 |