The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000795.3 | NP_000786.1 | 3 - 4 | 626 | 87 |
NM_016574.3 | NP_057658.2 | 3 - 4 | 626 | 87 | |
XM_017017296.1 | XP_016872785.1 | 3 - 4 | 608 | 87 | |
GenBank mRNA | AB209832.1 | - | 2 - 3 | 421 | 87 |
AB451480.1 | - | 2 - 3 | 391 | 87 | |
AF176812.1 | - | 2 - 3 | 400 | 87 | |
AK308777.1 | - | 3 - 4 | 456 | 87 | |
BC021195.2 | - | 3 - 4 | 486 | 87 | |
M29066.1 | - | 2 - 3 | 424 | 87 | |
M30625.1 | - | 3 - 4 | 727 | 87 | |
S62137.1 | - | 2 - 3 | 708 | 87 | |
S69899.1 | - | 3 - 4 | 490 | 87 | |
X51362.1 | - | 3 - 4 | 556 | 87 | |
X51645.1 | - | 2 - 3 | 419 | 87 |