The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276453.1 | NP_001263382.1 | 10 - 11 | 1492 | 71 |
NM_002265.5 | NP_002256.2 | 11 - 12 | 1817 | 71 | |
GenBank mRNA | AK299145.1 | - | 6 - 7 | 849 | 71 |
AK301463.1 | - | 10 - 11 | 1492 | 71 | |
AK314780.1 | - | 11 - 12 | 1695 | 71 | |
AK316421.1 | - | 10 - 11 | 1713 | 71 | |
BC003572.1 | - | 11 - 12 | 1691 | 71 | |
BC024045.1 | - | 11 - 12 | 1691 | 71 | |
BC036703.2 | - | 11 - 12 | 1717 | 71 | |
BT009797.1 | - | 11 - 12 | 1410 | 71 | |
L38951.1 | - | 11 - 12 | 1747 | 71 |