The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031217.3 | NP_112494.3 | 3 - 4 | 664 | 154 |
XM_017018379.1 | XP_016873868.1 | 3 - 4 | 1220 | 154 | |
GenBank mRNA | AB062483.1 | - | 3 - 4 | 565 | 154 |
AK312576.1 | - | 3 - 4 | 609 | 154 | |
AL136819.1 | - | 3 - 4 | 611 | 154 | |
AY791349.1 | - | 3 - 4 | 594 | 154 | |
BC026090.1 | - | 3 - 4 | 644 | 154 | |
BC048347.2 | - | 2 - 3 | 529 | 154 |