Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EOMES
Entrez Gene ID
8320
Gene Name
eomesodermin
Gene Aliases
TBR2
Chromosome Location
Chr.3: 27715949 - 27722715 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005442.3 NP_005433.2 5 - 6 1803 112
GenBank mRNA AB031038.1 - 5 - 6 1382 112
BC025363.1 - 4 - 5 496 112

View Full Product Details