Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRC2B
Entrez Gene ID
84726
Gene Name
proline rich coiled-coil 2B
Gene Aliases
BAT2L, BAT2L1, KIAA0515, LQFBS-1
Chromosome Location
Chr.9: 131430090 - 131500191 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013318.3 NP_037450.2 29 - 30 6532 119
GenBank mRNA AB011087.1 - 13 - 14 1801 119
AK026162.1 - 6 - 7 835 119
AK304849.1 - 2 - 3 269 119
BC012289.1 - 7 - 8 1024 119
BX647528.1 - 16 - 17 4171 119
BX647842.1 - 15 - 16 2235 119
CR749818.1 - 24 - 25 5607 119

View Full Product Details