Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRC2B
Entrez Gene ID
84726
Gene Name
proline rich coiled-coil 2B
Gene Aliases
BAT2L, BAT2L1, KIAA0515, LQFBS-1
Chromosome Location
Chr.9: 131430090 - 131500191 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013318.3 NP_037450.2 2 - 3 355 62
GenBank mRNA BC002872.2 - 3 - 4 406 62

View Full Product Details