The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037126.1 | NP_001032203.1 | 4 - 5 | 690 | 97 |
NM_021807.3 | NP_068579.3 | 4 - 5 | 690 | 97 | |
XM_005250523.4 | XP_005250580.1 | 4 - 5 | 674 | 97 | |
XM_017012495.1 | XP_016867984.1 | 4 - 5 | 674 | 97 | |
GenBank mRNA | AB051486.1 | - | 4 - 5 | 632 | 97 |
AF380839.1 | - | 4 - 5 | 687 | 97 | |
AK027688.1 | - | 4 - 5 | 684 | 97 | |
AK295404.1 | - | 4 - 5 | 698 | 97 | |
AL831989.2 | - | 4 - 5 | 685 | 97 | |
BC026174.1 | - | 4 - 5 | 666 | 97 | |
BC067263.1 | - | 4 - 5 | 667 | 97 |