Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRPK3
Entrez Gene ID
26576
Gene Name
SRSF protein kinase 3
Gene Aliases
MSSK-1, MSSK1, STK23
Chromosome Location
Chr.X: 153781001 - 153785732 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170760.1 NP_001164231.1 5 - 6 564 111
NM_001170761.1 NP_001164232.1 5 - 6 564 111
NM_014370.3 NP_055185.2 5 - 6 564 111
GenBank mRNA AF027406.1 - 5 - 6 502 111
AK301749.1 - 5 - 6 524 111
AK314828.1 - 5 - 6 524 111
BC092416.1 - 5 - 6 510 111
BC117124.1 - 5 - 6 558 111
DQ099381.1 - 5 - 6 478 111
U82808.1 - 5 - 6 382 111

View Full Product Details