Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAP47
Entrez Gene ID
116841
Gene Name
synaptosome associated protein 47
Gene Aliases
C1orf142, ESFI5812, HEL-S-290, HEL170, SNAP-47, SVAP1
Chromosome Location
Chr.1: 227728168 - 227781231 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323930.1 NP_001310859.1 1 - 2 269 55

View Full Product Details