Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
H2AFY
Entrez Gene ID
9555
Gene Name
H2A histone family member Y
Gene Aliases
H2A.y, H2A/y, H2AF12M, MACROH2A1.1, mH2A1, macroH2A1.2
Chromosome Location
Chr.5: 135334381 - 135399887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040158.1 NP_001035248.1 7 - 8 983 131
NM_004893.2 NP_004884.1 7 - 8 946 131
NM_138609.2 NP_613075.1 7 - 8 977 131
NM_138610.2 NP_613258.2 7 - 8 986 131
XM_005272132.1 XP_005272189.1 7 - 8 1352 131
XM_011543729.2 XP_011542031.1 7 - 8 903 131
XM_011543730.2 XP_011542032.1 7 - 8 903 131
GenBank mRNA AB209490.1 - 2 - 3 10044 131
AF041483.1 - 7 - 8 883 131
AF044286.1 - 6 - 7 769 131
AF054174.1 - 7 - 8 946 131
AK023409.1 - 7 - 8 977 131
AK123153.1 - 2 - 3 804 131
AK296014.1 - 3 - 4 461 131
AY134746.1 - 6 - 7 759 131
BC013331.1 - 7 - 8 912 131

View Full Product Details