Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EZH2
- Entrez Gene ID
-
2146
- Gene Name
- enhancer of zeste 2 polycomb repressive complex 2 subunit
- Gene Aliases
-
ENX-1, ENX1, EZH1, EZH2b, KMT6, KMT6A, WVS, WVS2
- Chromosome Location
-
Chr.7: 148807372 - 148884662 on Build GRCh38
View Full Product Details