The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170726.2 | NP_733844.1 | 15 - 16 | 2011 | 70 |
GenBank mRNA | U24267.1 | - | 15 - 16 | 1765 | 70 |