The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004914.3 | NP_004905.2 | 5 - 6 | 561 | 59 |
XM_005261859.4 | XP_005261916.1 | 6 - 7 | 645 | 59 | |
XM_006724381.3 | XP_006724444.1 | 6 - 7 | 648 | 59 | |
XM_006724382.3 | XP_006724445.1 | 5 - 6 | 573 | 59 | |
XM_011530544.2 | XP_011528846.1 | 6 - 7 | 653 | 59 | |
XM_011530545.2 | XP_011528847.1 | 6 - 7 | 649 | 59 | |
XM_011530546.2 | XP_011528848.1 | 6 - 7 | 655 | 59 | |
XM_011530547.2 | XP_011528849.1 | 4 - 5 | 495 | 59 | |
XM_011530550.2 | XP_011528852.1 | 6 - 7 | 659 | 59 | |
XM_017029104.1 | XP_016884593.1 | 5 - 6 | 569 | 59 | |
XM_017029105.1 | XP_016884594.1 | 4 - 5 | 491 | 59 | |
GenBank mRNA | AB023061.1 | - | 5 - 6 | 561 | 59 |
AF133588.1 | - | 5 - 6 | 531 | 59 | |
AY336745.1 | - | 4 - 5 | 455 | 59 | |
BC104989.1 | - | 5 - 6 | 550 | 59 | |
BC104991.1 | - | 5 - 6 | 550 | 59 | |
CR456553.1 | - | 5 - 6 | 539 | 59 |