Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EFNB3
Entrez Gene ID
1949
Gene Name
ephrin B3
Gene Aliases
EFL6, EPLG8, LERK8
Chromosome Location
Chr.17: 7705202 - 7711375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001406.3 NP_001397.1 2 - 3 813 103
GenBank mRNA AK314841.1 - 2 - 3 919 103
BC022499.1 - 1 - 2 252 103
BC042944.2 - 2 - 3 813 103
U57001.1 - 2 - 3 813 103
U62775.1 - 2 - 3 617 103
U66406.1 - 2 - 3 1029 103

View Full Product Details