The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303484.1 | NP_001290413.1 | 6 - 7 | 882 | 56 |
NM_031481.2 | NP_113669.1 | 7 - 8 | 945 | 56 | |
XM_011546149.2 | XP_011544451.1 | 7 - 8 | 1874 | 56 | |
XM_011546150.2 | XP_011544452.1 | 7 - 8 | 1744 | 56 | |
XM_011546151.2 | XP_011544453.1 | 7 - 8 | 1133 | 56 | |
XM_011546152.1 | XP_011544454.1 | 7 - 8 | 760 | 56 | |
XM_011546153.2 | XP_011544455.1 | 7 - 8 | 1755 | 56 | |
XM_011546154.2 | XP_011544456.1 | 7 - 8 | 1002 | 56 | |
XM_017028968.1 | XP_016884457.1 | 7 - 8 | 628 | 56 | |
XM_017028969.1 | XP_016884458.1 | 7 - 8 | 2774 | 56 | |
XM_017028970.1 | XP_016884459.1 | 8 - 9 | 2265 | 56 | |
XM_017028971.1 | XP_016884460.1 | 8 - 9 | 2254 | 56 | |
XM_017028972.1 | XP_016884461.1 | 8 - 9 | 2177 | 56 | |
XM_017028973.1 | XP_016884462.1 | 7 - 8 | 2202 | 56 | |
XM_017028974.1 | XP_016884463.1 | 8 - 9 | 2133 | 56 | |
XM_017028975.1 | XP_016884464.1 | 8 - 9 | 2045 | 56 | |
GenBank mRNA | AJ428203.1 | - | 7 - 8 | 901 | 56 |
AK309110.1 | - | 6 - 7 | 729 | 56 | |
AY008285.1 | - | 7 - 8 | 901 | 56 | |
BC031644.1 | - | 6 - 7 | 837 | 56 | |
CR456578.1 | - | 5 - 6 | 531 | 56 |