Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX22
Entrez Gene ID
79856
Gene Name
sorting nexin 22
Gene Aliases
-
Chromosome Location
Chr.15: 64151698 - 64157481 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024798.2 NP_079074.2 1 - 2 134 77
NR_073534.1 1 - 2 134 77
GenBank mRNA AK024014.1 - 1 - 2 134 77
AY044653.1 - 1 - 2 75 77
BC019655.1 - 1 - 2 112 77

View Full Product Details