The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281467.1 | NP_001268396.1 | 1 - 2 | 277 | 91 |
NM_015939.4 | NP_057023.2 | 1 - 2 | 277 | 91 | |
XM_017027875.1 | XP_016883364.1 | 1 - 2 | 252 | 91 | |
GenBank mRNA | AB032979.1 | - | 1 - 2 | 196 | 91 |
AF132943.1 | - | 1 - 2 | 230 | 91 | |
AK000613.1 | - | 1 - 2 | 203 | 91 | |
AK300812.1 | - | 1 - 2 | 124 | 91 | |
AK308648.1 | - | 1 - 2 | 217 | 91 | |
AK309205.1 | - | 1 - 2 | 254 | 91 | |
BC001262.1 | - | 1 - 2 | 196 | 91 | |
DC348968.1 | - | 1 - 2 | 277 | 91 |