Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZP3
Entrez Gene ID
7784
Gene Name
zona pellucida glycoprotein 3 (sperm receptor)
Gene Aliases
ZP3A, ZP3B, ZPC, Zp-3
Chromosome Location
Chr.7: 76397524 - 76442071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110354.1 NP_001103824.1 2 - 3 446 59
NM_007155.5 NP_009086.4 3 - 4 623 59
XM_017012586.1 XP_016868075.1 3 - 4 592 59
GenBank mRNA AK056788.1 - 1 - 2 1000 59
AK292763.1 - 3 - 4 623 59
BC113949.1 - 3 - 4 596 59
BI828655.1 - 3 - 4 596 59
M60504.1 - 2 - 3 446 59

View Full Product Details