Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZP3
Entrez Gene ID
7784
Gene Name
zona pellucida glycoprotein 3 (sperm receptor)
Gene Aliases
ZP3A, ZP3B, ZPC, Zp-3
Chromosome Location
Chr.7: 76397524 - 76442071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110354.1 NP_001103824.1 4 - 5 723 65
NM_007155.5 NP_009086.4 5 - 6 900 65
XM_017012586.1 XP_016868075.1 5 - 6 869 65
GenBank mRNA AK056788.1 - 3 - 4 1277 65
AK292763.1 - 5 - 6 900 65
BC113949.1 - 5 - 6 873 65
M60504.1 - 4 - 5 723 65

View Full Product Details