The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111020.2 | NP_001104490.1 | 6 - 7 | 567 | 86 |
NM_001130824.1 | NP_001124296.1 | 6 - 7 | 492 | 86 | |
NM_001130825.1 | NP_001124297.1 | 5 - 6 | 555 | 86 | |
NM_001319990.1 | NP_001306919.1 | 6 - 7 | 489 | 86 | |
NM_001319991.1 | NP_001306920.1 | 5 - 6 | 477 | 86 | |
NM_003169.3 | NP_003160.2 | 5 - 6 | 631 | 86 | |
XM_017027174.1 | XP_016882663.1 | 1 - 2 | 69 | 86 | |
GenBank mRNA | AB000516.1 | - | 6 - 7 | 541 | 86 |
AB209257.1 | - | 4 - 5 | 475 | 86 | |
AF040253.1 | - | 5 - 6 | 595 | 86 | |
AK302954.1 | - | 7 - 8 | 465 | 86 | |
AK303480.1 | - | 5 - 6 | 346 | 86 | |
BC024203.2 | - | 6 - 7 | 424 | 86 | |
U56402.1 | - | 6 - 7 | 436 | 86 | |
Y12790.1 | - | 6 - 7 | 531 | 86 |