The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001372.3 | NP_001363.2 | 61 - 62 | 11820 | 80 |
NM_004662.2 | NP_004653.2 | 7 - 8 | 1217 | 80 | |
XM_011523703.1 | XP_011522005.1 | 61 - 62 | 11784 | 80 | |
XM_017024292.1 | XP_016879781.1 | 60 - 61 | 11667 | 80 | |
XM_017024293.1 | XP_016879782.1 | 50 - 51 | 9728 | 80 | |
GenBank mRNA | AB002355.2 | - | 42 - 43 | 7498 | 80 |
AB290158.1 | - | 49 - 50 | 9637 | 80 | |
AF015265.1 | - | 2 - 3 | 265 | 80 | |
AF257737.1 | - | 61 - 62 | 11810 | 80 | |
AJ404468.1 | - | 61 - 62 | 11820 | 80 | |
AK302009.1 | - | 6 - 7 | 1033 | 80 | |
BC128261.1 | - | 5 - 6 | 757 | 80 | |
BC128421.1 | - | 5 - 6 | 757 | 80 | |
BC153883.1 | - | 42 - 43 | 7498 | 80 | |
X99947.1 | - | 7 - 8 | 1217 | 80 |