Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAH9
Entrez Gene ID
1770
Gene Name
dynein axonemal heavy chain 9
Gene Aliases
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20
Chromosome Location
Chr.17: 11598431 - 11970168 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372.3 NP_001363.2 16 - 17 2998 74
XM_011523703.1 XP_011522005.1 16 - 17 2962 74
XM_017024292.1 XP_016879781.1 15 - 16 2845 74
XM_017024293.1 XP_016879782.1 5 - 6 906 74
XM_017024294.1 XP_016879783.1 16 - 17 2962 74
XM_017024295.1 XP_016879784.1 16 - 17 2962 74
GenBank mRNA AB290158.1 - 4 - 5 815 74
AF257737.1 - 16 - 17 2988 74
AJ404468.1 - 16 - 17 2998 74

View Full Product Details