The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001372.3 | NP_001363.2 | 28 - 29 | 5879 | 93 |
XM_011523703.1 | XP_011522005.1 | 28 - 29 | 5843 | 93 | |
XM_017024292.1 | XP_016879781.1 | 27 - 28 | 5726 | 93 | |
XM_017024293.1 | XP_016879782.1 | 17 - 18 | 3787 | 93 | |
XM_017024294.1 | XP_016879783.1 | 28 - 29 | 5843 | 93 | |
XM_017024295.1 | XP_016879784.1 | 28 - 29 | 5843 | 93 | |
GenBank mRNA | AB002355.2 | - | 9 - 10 | 1557 | 93 |
AB290158.1 | - | 16 - 17 | 3696 | 93 | |
AF257737.1 | - | 28 - 29 | 5869 | 93 | |
AJ132088.1 | - | 2 - 3 | 192 | 93 | |
AJ404468.1 | - | 28 - 29 | 5879 | 93 | |
BC153883.1 | - | 9 - 10 | 1557 | 93 |