Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35C1
Entrez Gene ID
55343
Gene Name
solute carrier family 35 member C1
Gene Aliases
CDG2C, FUCT1
Chromosome Location
Chr.11: 45804072 - 45813016 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011520202.2 XP_011518504.1 1 - 2 132 87

View Full Product Details