The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324183.1 | NP_001311112.1 | 1 - 2 | 351 | 132 |
NM_001324184.1 | NP_001311113.1 | 1 - 2 | 351 | 132 | |
NM_001324185.1 | NP_001311114.1 | 1 - 2 | 351 | 132 | |
NR_136711.1 | 1 - 2 | 351 | 132 | ||
XM_011521891.2 | XP_011520193.1 | 1 - 2 | 456 | 132 | |
XM_011521892.2 | XP_011520194.1 | 1 - 2 | 456 | 132 |