The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013318.3 | NP_037450.2 | 13 - 14 | 2159 | 118 |
GenBank mRNA | AK309879.1 | - | 6 - 7 | 1105 | 118 |
CN369737.1 | - | 1 - 2 | 85 | 118 | |
CR749818.1 | - | 8 - 9 | 1234 | 118 | |
CR933661.1 | - | 1 - 2 | 46 | 118 |