The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133486.2 | NP_597846.1 | 4 - 5 | 854 | 97 |
GenBank mRNA | AB077699.1 | - | 4 - 5 | 854 | 97 |
AF467070.1 | - | 4 - 5 | 854 | 97 | |
AJ427919.1 | - | 4 - 5 | 854 | 97 |