The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136158.1 | NP_001129630.1 | 9 - 10 | 1797 | 75 |
XM_005278039.3 | XP_005278096.1 | 9 - 10 | 2565 | 75 | |
XM_006724537.2 | XP_006724600.1 | 9 - 10 | 2562 | 75 | |
XM_017029628.1 | XP_016885117.1 | 9 - 10 | 2550 | 75 | |
GenBank mRNA | BC011738.2 | - | 9 - 10 | 1353 | 75 |