Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5M
Entrez Gene ID
56953
Gene Name
5',3'-nucleotidase, mitochondrial
Gene Aliases
dNT-2, dNT2, mdN
Chromosome Location
Chr.17: 17303366 - 17347663 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020201.3 NP_064586.1 4 - 5 731 138
GenBank mRNA AF210652.1 - 4 - 5 762 138
BC035838.1 - 4 - 5 705 138

View Full Product Details