Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO15B
Entrez Gene ID
80022
Gene Name
myosin XVB
Gene Aliases
MYO15BP
Chromosome Location
Chr.17: 75587545 - 75626850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001309242.1 NP_001296171.1 20 - 21 4181 143
XM_017025120.1 XP_016880609.1 22 - 23 3888 143
XM_017025121.1 XP_016880610.1 22 - 23 3873 143
XM_017025122.1 XP_016880611.1 22 - 23 3852 143
XM_017025123.1 XP_016880612.1 22 - 23 3890 143
XM_017025124.1 XP_016880613.1 21 - 22 3762 143
XM_017025126.1 XP_016880615.1 17 - 18 3353 143
XM_017025127.1 XP_016880616.1 20 - 21 3196 143
XM_017025128.1 XP_016880617.1 21 - 22 2942 143
XM_017025129.1 XP_016880618.1 20 - 21 2174 143
XM_017025130.1 XP_016880619.1 19 - 20 2130 143
XM_017025131.1 XP_016880620.1 18 - 19 2569 143
XM_017025132.1 XP_016880621.1 17 - 18 2510 143
XM_017025133.1 XP_016880622.1 15 - 16 2035 143
XM_017025134.1 XP_016880623.1 16 - 17 2396 143
XM_017025135.1 XP_016880624.1 10 - 11 1052 143
XM_017025136.1 XP_016880625.1 10 - 11 1401 143
XM_017025137.1 XP_016880626.1 7 - 8 946 143
XM_017025141.1 XP_016880630.1 22 - 23 4154 143
XM_017025142.1 XP_016880631.1 22 - 23 4154 143
GenBank mRNA AB058686.1 - 10 - 11 2288 143
AF418286.1 - 18 - 19 4414 143
AF418290.1 - 15 - 16 4718 143

View Full Product Details