The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199096.1 | NP_001186025.1 | 18 - 19 | 1861 | 70 |
NM_001199097.1 | NP_001186026.1 | 19 - 20 | 1911 | 70 | |
NM_001199098.1 | NP_001186027.1 | 19 - 20 | 1842 | 70 | |
NM_001199099.1 | NP_001186028.1 | 19 - 20 | 1827 | 70 | |
NM_001286464.1 | NP_001273393.1 | 19 - 20 | 1962 | 70 | |
NM_003933.4 | NP_003924.2 | 19 - 20 | 2019 | 70 | |
XM_011522728.1 | XP_011521030.1 | 19 - 20 | 2565 | 70 | |
XM_011522729.1 | XP_011521031.1 | 19 - 20 | 2565 | 70 | |
XM_011522730.2 | XP_011521032.1 | 19 - 20 | 2565 | 70 | |
GenBank mRNA | AB018277.2 | - | 19 - 20 | 1871 | 70 |
AK294744.1 | - | 19 - 20 | 1787 | 70 | |
AK295645.1 | - | 19 - 20 | 1801 | 70 | |
AK299309.1 | - | 19 - 20 | 1871 | 70 | |
AK299358.1 | - | 18 - 19 | 1861 | 70 | |
AK315059.1 | - | 19 - 20 | 2019 | 70 | |
AL834321.1 | - | 19 - 20 | 1913 | 70 | |
BC104966.1 | - | 19 - 20 | 1882 | 70 | |
BC112129.1 | - | 19 - 20 | 1894 | 70 |