The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199096.1 | NP_001186025.1 | 22 - 23 | 2316 | 123 |
NM_001199097.1 | NP_001186026.1 | 23 - 24 | 2366 | 123 | |
NM_001199098.1 | NP_001186027.1 | 23 - 24 | 2297 | 123 | |
NM_001199099.1 | NP_001186028.1 | 23 - 24 | 2282 | 123 | |
NM_001286464.1 | NP_001273393.1 | 23 - 24 | 2417 | 123 | |
NM_003933.4 | NP_003924.2 | 23 - 24 | 2474 | 123 | |
XM_011522728.1 | XP_011521030.1 | 23 - 24 | 3020 | 123 | |
XM_011522729.1 | XP_011521031.1 | 23 - 24 | 3020 | 123 | |
XM_011522730.2 | XP_011521032.1 | 23 - 24 | 3020 | 123 | |
GenBank mRNA | AB018277.2 | - | 23 - 24 | 2326 | 123 |
AK294744.1 | - | 23 - 24 | 2242 | 123 | |
AK295645.1 | - | 23 - 24 | 2256 | 123 | |
AK299309.1 | - | 23 - 24 | 2326 | 123 | |
AK299358.1 | - | 22 - 23 | 2316 | 123 | |
AK315059.1 | - | 23 - 24 | 2474 | 123 | |
AL834321.1 | - | 23 - 24 | 2368 | 123 | |
BC104966.1 | - | 23 - 24 | 2337 | 123 | |
BC112129.1 | - | 23 - 24 | 2349 | 123 |