Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAIAP3
Entrez Gene ID
8938
Gene Name
BAI1 associated protein 3
Gene Aliases
BAP3
Chromosome Location
Chr.16: 1333605 - 1349441 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199096.1 NP_001186025.1 25 - 26 2664 68
NM_001199097.1 NP_001186026.1 26 - 27 2714 68
NM_001199098.1 NP_001186027.1 26 - 27 2645 68
NM_001199099.1 NP_001186028.1 26 - 27 2630 68
NM_001286464.1 NP_001273393.1 26 - 27 2765 68
NM_003933.4 NP_003924.2 26 - 27 2822 68
XM_011522728.1 XP_011521030.1 26 - 27 3368 68
XM_011522729.1 XP_011521031.1 26 - 27 3368 68
XM_011522730.2 XP_011521032.1 26 - 27 3368 68
GenBank mRNA AB018277.2 - 26 - 27 2674 68
AK294744.1 - 26 - 27 2590 68
AK295645.1 - 26 - 27 2604 68
AK299309.1 - 26 - 27 2674 68
AK299358.1 - 25 - 26 2664 68
AK315059.1 - 26 - 27 2822 68
AL834321.1 - 26 - 27 2716 68
BC104966.1 - 26 - 27 2685 68
BC112129.1 - 26 - 27 2697 68

View Full Product Details