The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199096.1 | NP_001186025.1 | 27 - 28 | 2861 | 62 |
NM_001199097.1 | NP_001186026.1 | 28 - 29 | 2911 | 62 | |
NM_001199098.1 | NP_001186027.1 | 28 - 29 | 2842 | 62 | |
NM_001199099.1 | NP_001186028.1 | 28 - 29 | 2827 | 62 | |
NM_001286464.1 | NP_001273393.1 | 28 - 29 | 2962 | 62 | |
NM_003933.4 | NP_003924.2 | 28 - 29 | 3019 | 62 | |
XM_011522728.1 | XP_011521030.1 | 28 - 29 | 3565 | 62 | |
XM_011522729.1 | XP_011521031.1 | 28 - 29 | 3565 | 62 | |
XM_011522730.2 | XP_011521032.1 | 28 - 29 | 3565 | 62 | |
GenBank mRNA | AB018277.2 | - | 28 - 29 | 2871 | 62 |
AF038191.1 | - | 3 - 4 | 211 | 62 | |
AK294744.1 | - | 28 - 29 | 2787 | 62 | |
AK295645.1 | - | 28 - 29 | 2801 | 62 | |
AK299309.1 | - | 28 - 29 | 2871 | 62 | |
AK299358.1 | - | 27 - 28 | 2861 | 62 | |
AK315059.1 | - | 28 - 29 | 3019 | 62 | |
AL834321.1 | - | 28 - 29 | 2913 | 62 | |
BC104966.1 | - | 28 - 29 | 2882 | 62 | |
BC112129.1 | - | 28 - 29 | 2894 | 62 |