Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAIAP3
Entrez Gene ID
8938
Gene Name
BAI1 associated protein 3
Gene Aliases
BAP3
Chromosome Location
Chr.16: 1333605 - 1349441 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199096.1 NP_001186025.1 31 - 32 3260 87
NM_001199097.1 NP_001186026.1 32 - 33 3310 87
NM_001199098.1 NP_001186027.1 32 - 33 3241 87
NM_001199099.1 NP_001186028.1 32 - 33 3226 87
NM_001286464.1 NP_001273393.1 32 - 33 3361 87
NM_003933.4 NP_003924.2 32 - 33 3418 87
XM_011522728.1 XP_011521030.1 32 - 33 3964 87
XM_011522729.1 XP_011521031.1 32 - 33 3964 87
XM_011522730.2 XP_011521032.1 32 - 33 3964 87
GenBank mRNA AB018277.2 - 32 - 33 3270 87
AF038191.1 - 7 - 8 610 87
AK294744.1 - 32 - 33 3186 87
AK295645.1 - 32 - 33 3200 87
AK299309.1 - 32 - 33 3270 87
AK299358.1 - 31 - 32 3260 87
AK315059.1 - 32 - 33 3418 87
AL834321.1 - 32 - 33 3312 87
BC104966.1 - 32 - 33 3281 87
BC112129.1 - 32 - 33 3293 87

View Full Product Details