Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPHP1
Entrez Gene ID
4867
Gene Name
nephrocystin 1
Gene Aliases
JBTS4, NPH1, SLSN1
Chromosome Location
Chr.2: 110123336 - 110205062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000272.3 NP_000263.2 9 - 10 1119 75
NM_001128178.1 NP_001121650.1 9 - 10 951 75
NM_001128179.1 NP_001121651.1 7 - 8 762 75
NM_207181.2 NP_997064.2 9 - 10 1116 75
XM_005263676.1 XP_005263733.1 9 - 10 900 75
XM_005263677.1 XP_005263734.1 9 - 10 897 75
XM_005263678.2 XP_005263735.1 9 - 10 1068 75
XM_005263679.1 XP_005263736.1 9 - 10 897 75
XM_006712551.1 XP_006712614.1 9 - 10 1068 75
XM_006712552.2 XP_006712615.1 9 - 10 1068 75
XM_011511244.1 XP_011509546.1 9 - 10 1068 75
XM_017004218.1 XP_016859707.1 9 - 10 900 75
GenBank mRNA AF023674.1 - 9 - 10 1020 75
AJ001815.1 - 9 - 10 1055 75
AK225600.1 - 9 - 10 881 75
AK299011.1 - 9 - 10 877 75
AK307654.1 - 9 - 10 881 75
BC062574.1 - 7 - 8 672 75
BX571745.1 - 8 - 9 2460 75

View Full Product Details