The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021120.3 | NP_066943.2 | 5 - 6 | 1182 | 64 |
XM_006724625.2 | XP_006724688.1 | 5 - 6 | 1187 | 64 | |
XM_006724626.2 | XP_006724689.1 | 5 - 6 | 1187 | 64 | |
XM_011530883.1 | XP_011529185.1 | 5 - 6 | 1187 | 64 | |
GenBank mRNA | AB209328.1 | - | 5 - 6 | 1182 | 64 |
BC093864.1 | - | 5 - 6 | 883 | 64 | |
BC093866.1 | - | 5 - 6 | 883 | 64 | |
U49089.1 | - | 5 - 6 | 1068 | 64 |