The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166278.1 | NP_001159750.1 | 5 - 6 | 637 | 59 |
NM_020730.2 | NP_065781.1 | 7 - 8 | 1180 | 59 | |
XM_005262248.3 | XP_005262305.1 | 7 - 8 | 777 | 59 | |
XM_006724625.2 | XP_006724688.1 | 13 - 14 | 2226 | 59 | |
XM_017029325.1 | XP_016884814.1 | 5 - 6 | 2487 | 59 | |
XM_017029326.1 | XP_016884815.1 | 5 - 6 | 566 | 59 | |
GenBank mRNA | AB033058.1 | - | 7 - 8 | 1178 | 59 |
AK303377.1 | - | 5 - 6 | 637 | 59 | |
AK304020.1 | - | 7 - 8 | 893 | 59 | |
AK316518.1 | - | 7 - 8 | 879 | 59 |