Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RADIL
Entrez Gene ID
55698
Gene Name
Rap associating with DIL domain
Gene Aliases
RASIP2
Chromosome Location
Chr.7: 4799109 - 4883704 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK296325.1 - 1 - 2 225 167

View Full Product Details