Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RADIL
Entrez Gene ID
55698
Gene Name
Rap associating with DIL domain
Gene Aliases
RASIP2
Chromosome Location
Chr.7: 4799109 - 4883704 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018059.4 NP_060529.4 10 - 11 2467 144
GenBank mRNA AB058752.1 - 10 - 11 2411 144
AK000997.1 - 6 - 7 905 144
AK001186.1 - 7 - 8 1450 144
AK294745.1 - 9 - 10 2266 144
AK296325.1 - 9 - 10 1988 144
AK302549.1 - - 2459 144
AL136731.1 - 5 - 6 698 144
BC117317.1 - 9 - 10 2353 144
BC126311.1 - 9 - 10 2353 144
BC143526.1 - 9 - 10 2353 144
EF560723.1 - 9 - 10 2376 144

View Full Product Details