The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000548.4 | NP_000539.2 | 30 - 31 | 3815 | 69 |
NM_001077183.2 | NP_001070651.1 | 29 - 30 | 3683 | 69 | |
NM_001114382.2 | NP_001107854.1 | 30 - 31 | 3815 | 69 | |
NM_001318827.1 | NP_001305756.1 | 28 - 29 | 3575 | 69 | |
NM_001318829.1 | NP_001305758.1 | 28 - 29 | 3519 | 69 | |
NM_001318831.1 | NP_001305760.1 | 26 - 27 | 3309 | 69 | |
NM_001318832.1 | NP_001305761.1 | 29 - 30 | 3584 | 69 | |
XM_005255529.4 | XP_005255586.2 | 29 - 30 | 3573 | 69 | |
XM_005255531.4 | XP_005255588.2 | 29 - 30 | 3573 | 69 | |
XM_011522636.2 | XP_011520938.1 | 30 - 31 | 3702 | 69 | |
XM_011522637.2 | XP_011520939.1 | 30 - 31 | 3699 | 69 | |
XM_011522638.2 | XP_011520940.2 | 29 - 30 | 3795 | 69 | |
XM_011522639.2 | XP_011520941.1 | 29 - 30 | 3573 | 69 | |
XM_011522640.2 | XP_011520942.1 | 29 - 30 | 3570 | 69 | |
XM_017023615.1 | XP_016879104.1 | 30 - 31 | 3699 | 69 | |
XM_017023616.1 | XP_016879105.1 | 29 - 30 | 3570 | 69 | |
XM_017023617.1 | XP_016879106.1 | 28 - 29 | 3666 | 69 | |
XM_017023618.1 | XP_016879107.1 | 31 - 32 | 3789 | 69 | |
GenBank mRNA | AB210000.1 | - | 29 - 30 | 3589 | 69 |
AK125096.1 | - | 4 - 5 | 1442 | 69 | |
AK295672.1 | - | 28 - 29 | 3406 | 69 | |
AK295728.1 | - | 29 - 30 | 3584 | 69 | |
AK299343.1 | - | 28 - 29 | 3456 | 69 | |
BC025364.1 | - | 4 - 5 | 603 | 69 | |
BC046929.1 | - | 4 - 5 | 603 | 69 | |
BC150300.1 | - | 30 - 31 | 3690 | 69 | |
BX647816.1 | - | 26 - 27 | 3172 | 69 | |
KJ534954.1 | - | 28 - 29 | 3486 | 69 | |
KJ534955.1 | - | 28 - 29 | 3483 | 69 | |
KJ534958.1 | - | 28 - 29 | 3483 | 69 | |
KJ535091.1 | - | 28 - 29 | 3486 | 69 | |
KJ535094.1 | - | 14 - 15 | 1801 | 69 | |
X75621.1 | - | 29 - 30 | 3633 | 69 |