The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139057.3 | NP_620688.2 | 9 - 10 | 1432 | 65 |
XM_005254872.3 | XP_005254929.1 | 9 - 10 | 2348 | 65 | |
XM_011521312.2 | XP_011519614.1 | 9 - 10 | 2348 | 65 | |
XM_017021973.1 | XP_016877462.1 | 10 - 11 | 2481 | 65 | |
XM_017021974.1 | XP_016877463.1 | 10 - 11 | 2483 | 65 | |
XM_017021975.1 | XP_016877464.1 | 10 - 11 | 2481 | 65 | |
XM_017021976.1 | XP_016877465.1 | 9 - 10 | 1193 | 65 | |
XM_017021977.1 | XP_016877466.1 | 10 - 11 | 2480 | 65 | |
XM_017021978.1 | XP_016877467.1 | 3 - 4 | 3739 | 65 | |
XM_017021979.1 | XP_016877468.1 | 2 - 3 | 189 | 65 | |
XM_017021980.1 | XP_016877469.1 | 2 - 3 | 399 | 65 | |
XM_017021981.1 | XP_016877470.1 | 10 - 11 | 2478 | 65 | |
XM_017021984.1 | XP_016877473.1 | 8 - 9 | 1007 | 65 | |
GenBank mRNA | AJ315735.1 | - | 9 - 10 | 1349 | 65 |
AK131344.1 | - | 8 - 9 | 978 | 65 |