Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTS17
Entrez Gene ID
170691
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 17
Gene Aliases
-
Chromosome Location
Chr.15: 99971438 - 100342924 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139057.3 NP_620688.2 1 - 2 185 74
XM_005254872.3 XP_005254929.1 1 - 2 1101 74
XM_011521312.2 XP_011519614.1 1 - 2 1101 74
XM_017021973.1 XP_016877462.1 1 - 2 1102 74
XM_017021974.1 XP_016877463.1 1 - 2 1104 74
XM_017021975.1 XP_016877464.1 1 - 2 1102 74
XM_017021977.1 XP_016877466.1 1 - 2 1101 74
XM_017021981.1 XP_016877470.1 1 - 2 1099 74
GenBank mRNA AJ315735.1 - 1 - 2 102 74

View Full Product Details