Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP53
Entrez Gene ID
54532
Gene Name
ubiquitin specific peptidase 53
Gene Aliases
-
Chromosome Location
Chr.4: 119212583 - 119295518 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019050.2 NP_061923.2 15 - 16 3242 109
XM_005263073.3 XP_005263130.1 14 - 15 3116 109
XM_005263075.2 XP_005263132.1 14 - 15 3131 109
XM_005263076.2 XP_005263133.1 14 - 15 3128 109
XM_005263077.3 XP_005263134.1 15 - 16 3281 109
XM_006714240.1 XP_006714303.1 13 - 14 2317 109
XM_011532037.2 XP_011530339.1 15 - 16 3294 109
XM_011532038.2 XP_011530340.1 14 - 15 2571 109
XM_017008312.1 XP_016863801.1 16 - 17 3408 109
XM_017008313.1 XP_016863802.1 15 - 16 3093 109
XM_017008314.1 XP_016863803.1 15 - 16 3092 109
XM_017008315.1 XP_016863804.1 15 - 16 3162 109
XM_017008316.1 XP_016863805.1 13 - 14 2980 109
XM_017008317.1 XP_016863806.1 14 - 15 3105 109
XM_017008318.1 XP_016863807.1 15 - 16 3227 109
GenBank mRNA AB037771.1 - 8 - 9 1691 109
AJ583824.1 - 12 - 13 2173 109
BX648339.1 - 13 - 14 2330 109
CR749490.1 - 16 - 17 3358 109

View Full Product Details