The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015726.3 | NP_056541.2 | 11 - 12 | 1650 | 105 |
NR_028103.1 | 11 - 12 | 1950 | 105 | ||
NR_028104.1 | 10 - 11 | 1876 | 105 | ||
GenBank mRNA | AK026481.1 | - | 11 - 12 | 1716 | 105 |
AK223475.1 | - | 10 - 11 | 1505 | 105 | |
AK303188.1 | - | 12 - 13 | 1518 | 105 | |
AK312690.1 | - | 10 - 11 | 1799 | 105 | |
BC098271.1 | - | 11 - 12 | 1556 | 105 | |
BC099709.1 | - | 11 - 12 | 1556 | 105 | |
BC099846.1 | - | 11 - 12 | 1556 | 105 | |
CR749801.1 | - | 11 - 12 | 1873 | 105 | |
U06631.1 | - | 11 - 12 | 1614 | 105 |