The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013836.1 | NP_001013858.1 | 4 - 5 | 559 | 109 |
NM_001013837.1 | NP_001013859.1 | 4 - 5 | 556 | 109 | |
NM_001304523.1 | NP_001291452.1 | 3 - 4 | 380 | 109 | |
NM_003550.2 | NP_003541.2 | 4 - 5 | 596 | 109 | |
XM_011515567.1 | XP_011513869.1 | 2 - 3 | 303 | 109 | |
XM_011515568.2 | XP_011513870.1 | 2 - 3 | 303 | 109 | |
XM_017012690.1 | XP_016868179.1 | 4 - 5 | 774 | 109 | |
GenBank mRNA | AF083811.1 | - | 4 - 5 | 542 | 109 |
AF123318.1 | - | 2 - 3 | 293 | 109 | |
AK310831.1 | - | 4 - 5 | 556 | 109 | |
AK314227.1 | - | 3 - 4 | 377 | 109 | |
BC009964.2 | - | 3 - 4 | 506 | 109 | |
BQ684437.1 | - | 4 - 5 | 542 | 109 | |
U33822.1 | - | 3 - 4 | 472 | 109 |