Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BORCS8-MEF2B
Entrez Gene ID
4207
Gene Name
BORCS8-MEF2B readthrough
Gene Aliases
LOC729991-MEF2B, MEF2B, MEF2BNB-MEF2B
Chromosome Location
Chr.19: 19145567 - 19192591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005919.3 NP_005910.1 6 - 7 1119 101
NR_027307.1 8 - 9 1297 101
NR_027308.1 8 - 9 1297 101
GenBank mRNA AK301086.1 - 8 - 9 864 101
AK316328.1 - 8 - 9 864 101
BC126245.1 - 5 - 6 542 101
BC136457.1 - 4 - 5 466 101
BC171767.1 - 4 - 5 466 101
X63380.1 - 8 - 9 834 101
X68502.1 - 6 - 7 641 101

Gene Symbol
MEF2B
Entrez Gene ID
100271849
Gene Name
myocyte enhancer factor 2B
Gene Aliases
RSRFR2
Chromosome Location
Chr.19: 19145567 - 19170289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145785.1 NP_001139257.1 4 - 5 506 101
GenBank mRNA AK057210.1 - 4 - 5 489 101

View Full Product Details